missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals EPPS, 0.2M buffer soln., pH 8.0

Catalog No. AAJ61296AE
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61296AE 100 mL
AAJ61296AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61296AE Supplier Thermo Scientific Chemicals Supplier No. J61296AE

CAS: 16052-06-5 | C9H20N2O4S | 252.329 g/mol

EPPS or HEPPS is a commonly used Good’s buffer. It is often used as separator in ultrathin isoelectric focusing gels and enhances the resolution of phosphoglucomutase. It is used as a buffering agent in biology and biochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
EPPS or HEPPS is a commonly used Good’s buffer. It is often used as separator in ultrathin isoelectric focusing gels and enhances the resolution of phosphoglucomutase. It is used as a buffering agent in biology and biochemistry.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

Molecular Formula C9H20N2O4S
Molecular Weight (g/mol) 252.329
MDL Number MFCD00006160
InChI Key OWXMKDGYPWMGEB-UHFFFAOYSA-N
Synonym HEPPS
PubChem CID 85255
ChEBI CHEBI:42298
IUPAC Name 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CCCS(=O)(=O)O)CCO

Specifications

Concentration 0.2 M
pH 8.0
Physical Form Liquid
Quantity 100 mL
Solubility Information It is soluble in water.
Chemical Name or Material EPPS buffer soln.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only