Learn More
Eppendorf™ Rotor for Centrifuge 5427 R
Eppendorf Rotor for Centrifuge 5427 R redefine efficiency. Tailor your centrifugation with various capacities and speeds. Explore accessory options for enhanced versatility, ensuring precise sample processing for diverse applications.
$1,734.69 - $3,807.46
| Catalog Number | Mfr. No. | Capacity | Includes | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity | Includes | Price | Quantity & Availability | ||||
|
05100157
|
Eppendorf™
5409702009 |
24 x 1.5 to 2.0 mL Tubes | Aerosol-tight with Rotor Lid |
Each for $2,438.25
|
|
||||
|
05100158
|
Eppendorf™
5409706004 |
30 x 1.5 to 2.0 mL Tubes | Aerosol-tight with Rotor Lid |
Each for $2,438.25
|
|
||||
|
05100159
|
Eppendorf™
5409710001 |
48 x 1.5 to 2.0 mL Tubes | Aerosol-tight with Rotor Lid |
Each for $2,821.15
|
|
||||
|
05100160
|
Eppendorf™
5409700006 |
12 x 5 mL Eppendorf Tubes | Aerosol-tight with Rotor Lid |
Each for $3,393.39
|
|
||||
|
05100161
|
Eppendorf™
5409704001 |
24 Spin Columns | Aerosol-tight with Rotor Lid |
Each for $2,943.19
|
|
||||
|
05100162
|
Eppendorf™
5409708007 |
30 x 1.5 to 2.0 mL Tubes | Rotor Lid |
Each for $1,734.69
|
|
||||
|
05414314
|
Eppendorf™
5409714007 |
48 x 0.2 mL PCR Tubes | Plastic Rotor Lid |
Each for $1,848.48
|
|
||||
|
05414315
|
Eppendorf™
5409715003 |
24 x 1.5/2.0 mL tubes | Aerosol-tight with Rotor Lid |
Each for $3,807.46
|
|
||||
Description
- Precision-engineered for optimal sample separation.
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
- Accessories (adaptors and buckets) further customize your centrifuge setup.
- Aerosol-tight options available.