Learn More
Eppendorf™ Rotor for Centrifuge 5427 R
Catalog No. 05414315
Change view
Click to view available options
Capacity:
12 x 5 mL Eppendorf Tubes
24 Spin Columns
24 x 1.5 to 2.0 mL Tubes
24 x 1.5/2.0 mL tubes
30 x 1.5 to 2.0 mL Tubes
48 x 1.5 to 2.0 mL Tubes
Includes:
Aerosol-tight with Rotor Lid
Rotor Lid
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Includes
05414315 24 x 1.5/2.0 mL tubes Aerosol-tight with Rotor Lid
05100157 24 x 1.5 to 2.0 mL Tubes Aerosol-tight with Rotor Lid
05100158 30 x 1.5 to 2.0 mL Tubes Aerosol-tight with Rotor Lid
05100159 48 x 1.5 to 2.0 mL Tubes Aerosol-tight with Rotor Lid
05100160 12 x 5 mL Eppendorf Tubes Aerosol-tight with Rotor Lid
05100161 24 Spin Columns Aerosol-tight with Rotor Lid
05100162 30 x 1.5 to 2.0 mL Tubes Rotor Lid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 05414315 Supplier Eppendorf™ Supplier No. 5409715003
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Eppendorf Rotor for Centrifuge 5427 R redefine efficiency. Tailor your centrifugation with various capacities and speeds. Explore accessory options for enhanced versatility, ensuring precise sample processing for diverse applications.
  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight options available.
Specifications
Type Rotor
Capacity 24 x 1.5/2.0 mL tubes
Max. RCF 16,049 x g
Max. Speed 12,700 rpm
For Use With (Equipment) Centrifuge 5427 R
Includes Aerosol-tight with Rotor Lid
Model S-24-11-AT