Learn More
Eppendorf™ Reference™ 2, Mechanical Single-Channel Pipettes Multi-Packs
Numéro de catalogue. p-4816407
Change view
Click to view available options
Volume (Metric):
0.1-2.5 μL, 0.5-10 μL, 10-100 μL, 100-1000 μL
0.1-2.5 μL, 0.5-10 μL, 2-20 μL, 10-100 μL, 20-200 μL, 100-1000 μL
0.1-2.5 μL, 2-20 μL, 20-200 μL, 100-1,000 μL
0.5-10 μL, 10-100 μL, 100-1000 μL
0.5-10 μL, 10-100 μL, 30-300 μL, 100-1000 μL, 0.5-5 mL, 1-10 mL
0.5-10 μL, 2-20 μL, 20-200 μL, 100-1000 μL
100-1000 μL, 0.5-5 mL, 1-10 mL
2-20 μL, 20-200 μL, 100-1000 μL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Volume (Metric)
05412524 0.1-2.5 μL, 2-20 μL, 20-200 μL, 100-1,000 μL
05412521 2-20 μL, 20-200 μL, 100-1000 μL
05412520 0.5-10 μL, 10-100 μL, 100-1000 μL
05412522 100-1000 μL, 0.5-5 mL, 1-10 mL
05412523 0.1-2.5 μL, 0.5-10 μL, 10-100 μL, 100-1000 μL
05412525 0.5-10 μL, 2-20 μL, 20-200 μL, 100-1000 μL
05412527 0.5-10 μL, 10-100 μL, 30-300 μL, 100-1000 μL, 0.5-5 mL, 1-10 mL
01671119 0.1-2.5 μL, 0.5-10 μL, 2-20 μL, 10-100 μL, 20-200 μL, 100-1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Numéro de catalogue. 05412524 Fournisseur Eppendorf™ Code fournisseur 2231302003
Il en reste null
Minimize risk of aerosol contamination and reduce thumb strain with these variable volume pipettes. Eppendorf™ Reference™ 2 Variable Volume Pipettes are ideal for pipetting and mixing liquids, filling of plates, gels and reaction vessels, and phase extraction and removal of supernatants. Available in packs of three, four and six.
  • Integrated pipetting system with internal RFID technology
  • Serial number is printed on multiple components of pipette
  • Color coded and volume labeled pipette for quick identification of volume size/tip size
  • Round upper part of pipette makes it possible to work in every position
  • Four-digit display for exact, quick and secure volume setting (volume lock included)
  • Spring-loaded tip cone reduces effort during operation/ejection
  • Sturdy upper handle, movable parts and external edges made from stainless-steel make pipette robust
  • High flexibility with ability to remove single cones
  • Ensures continuous identical pipet alignment during work process
  • Fully autoclavable pipet with smooth surface and few rough edges
  • Single-button operation
Spécifications
Autoclavable Autoclavable
Color Assorted
Number of Channels 1
Product Line Reference 2
Type 4-Pipette Multipack
Volume (Metric) 0.1-2.5 μL, 2-20 μL, 20-200 μL, 100-1,000 μL
Includes 4 Variable Volume Pipettes (Yellow)