Learn More
Eppendorf™ Reference™ 2, Mechanical Single-Channel Pipettes Multi-Packs
Minimize risk of aerosol contamination and reduce thumb strain with these variable volume pipettes. Eppendorf™ Reference™ 2 Variable Volume Pipettes are ideal for pipetting and mixing liquids, filling of plates, gels and reaction vessels, and phase extraction and removal of supernatants. Available in packs of three, four and six.
Supplier:  Eppendorf™ 2231302005
 View more versions of this product
Catalog No. 05412525
Only null left
Description
Description
  • Integrated pipetting system with internal RFID technology
  • Serial number is printed on multiple components of pipette
  • Color coded and volume labeled pipette for quick identification of volume size/tip size
  • Round upper part of pipette makes it possible to work in every position
  • Four-digit display for exact, quick and secure volume setting (volume lock included)
  • Spring-loaded tip cone reduces effort during operation/ejection
  • Sturdy upper handle, movable parts and external edges made from stainless-steel make pipette robust
  • High flexibility with ability to remove single cones
  • Ensures continuous identical pipet alignment during work process
  • Fully autoclavable pipet with smooth surface and few rough edges
  • Single-button operation
Specifications
Chemical Identifiers
Autoclavable
1
Specifications
Autoclavable
1
4-Pipette Multipack
4 Variable Volume Pipettes (Yellow)
Assorted
Reference 2
0.5-10 μL, 2-20 μL, 20-200 μL, 100-1000 μL
Product Suggestions
Product Suggestions