Learn More
Agilent BioTek Epoch 2 Microplate Spectrophotometer, IVD

The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. For In Vitro Diagnostic Use.
Supplier: Agilent BioTek EPOCH2TSCSI
Description
The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. Measurements can be performed in 6- to 384-well microplates, cuvets, and in microvolume samples with the available Take3 microvolume plate. The broad wavelength range enables applications from nucleic acid and protein quantification in the low UV to microbial growth assays higher wavelengths
Epoch 2 is easy to use and the optional touch screen interface makes it simple to choose predefined protocols or to define custom programs. Endpoint, kinetic, spectral scanning and well area scanning modes, plus incubation and shaking enable wide-ranging applications.
Chemical Identifiers
| Photodiode | |
| 96 wells: 8 sec., 384 wells: 14 sec. | |
| CE and TUV marked. RoHS compliant. | |
| 22 in. | |
| 200 to 999 nm, 1 nm increments |
| Xenon Flash Lamp | |
| ±2 nm | |
| Nucleic acid and protein quantification to ELISA and microbial growth assays | |
| 50/60 Hz |
Specifications
| Photodiode | |
| 96 wells: 8 sec., 384 wells: 14 sec. | |
| CE and TUV marked. RoHS compliant. | |
| 22 in. | |
| Touchscreen, Cuvette Port | |
| 200 to 999 nm, 1 nm increments |
| Xenon Flash Lamp | |
| ±2 nm | |
| Nucleic acid and protein quantification to ELISA and microbial growth assays | |
| 50/60 Hz | |
| EPOCH2TSC |