missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek Epoch 2 Microplate Spectrophotometer, IVD

Description
The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. Measurements can be performed in 6- to 384-well microplates, cuvets, and in microvolume samples with the available Take3 microvolume plate. The broad wavelength range enables applications from nucleic acid and protein quantification in the low UV to microbial growth assays higher wavelengths
Epoch 2 is easy to use and the optional touch screen interface makes it simple to choose predefined protocols or to define custom programs. Endpoint, kinetic, spectral scanning and well area scanning modes, plus incubation and shaking enable wide-ranging applications.
Specifications
Specifications
| Detector Type | Photodiode |
| Light Source | Xenon Flash Lamp |
| Measurement Speed | 96 wells: 8 sec., 384 wells: 14 sec. |
| Accuracy | ±2 nm |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| For Use With (Application) | Nucleic acid and protein quantification to ELISA and microbial growth assays |
| Height (English) | 22 in. |
| Hertz | 50/60 Hz |
| Includes | Touchscreen |
| Model | EPOCH2TS |
| Show More |