Learn More
Agilent BioTek Epoch 2 Microplate Spectrophotometer, IVD
GREENER_CHOICE
The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. For In Vitro Diagnostic Use.
Supplier:  Agilent BioTek EPOCH2NSCSI
 View more versions of this product
Catalog No. BTEPOCH2NCI
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. Measurements can be performed in 6- to 384-well microplates, cuvets, and in microvolume samples with the available Take3 microvolume plate. The broad wavelength range enables applications from nucleic acid and protein quantification in the low UV to microbial growth assays higher wavelengths
Epoch 2 is easy to use and the optional touch screen interface makes it simple to choose predefined protocols or to define custom programs. Endpoint, kinetic, spectral scanning and well area scanning modes, plus incubation and shaking enable wide-ranging applications.
Specifications
Chemical Identifiers
Photodiode
96 wells: 8 sec., 384 wells: 14 sec.
CE and TUV marked. RoHS compliant.
22 in.
200 to 999 nm, 1 nm increments
Xenon Flash Lamp
±2 nm
Nucleic acid and protein quantification to ELISA and microbial growth assays
50/60 Hz
Specifications
Photodiode
96 wells: 8 sec., 384 wells: 14 sec.
CE and TUV marked. RoHS compliant.
22 in.
Cuvette Port
200 to 999 nm, 1 nm increments
Xenon Flash Lamp
±2 nm
Nucleic acid and protein quantification to ELISA and microbial growth assays
50/60 Hz
EPOCH2NSC
Product Suggestions
Product Suggestions