missing translation for 'onlineSavingsMsg'
Learn More

Eosin Y TS, (U.S.P. Test Solution), Spectrum™ Chemical

Catalog No. 18602413
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602413 500 mL Amber Glass Bottle
18602414 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18602413 Supplier Spectrum Chemical Mfg Cor Supplier No. E146500ML
Get Up to $500 in Chemicals with Purchase
View all promotions

E-146, 15086-94-9

Spectrum™ Chemical Eosin Y TS, (U.S.P. Test Solution) is a solution, sometimes designated with 'TS or Test Solution', that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

Chemical Identifiers

CAS 17372-87-1 , 7732-18-5
Molecular Formula C20H6Br4Na2O5
Molecular Weight (g/mol) 691.86
InChI Key SEACYXSIPDVVMV-UHFFFAOYSA-L
IUPAC Name disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12

Specifications

CAS Max % 0.1%, 99.9%
Grade Chemical Solution
Packaging Amber Glass Bottle
Quantity 500 mL
Chemical Name or Material Eosin Y TS