missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eosin Y, Reagent, ACS, Spectrum™ Chemical
E1295, 17372-87-1, C20H6Br4Na2O5
$261.29 - $2,062.25
Chemical Identifiers
| CAS | 17372-87-1 |
|---|---|
| Molecular Formula | C20H6Br4Na2O5 |
| Molecular Weight (g/mol) | 691.86 |
| InChI Key | SEACYXSIPDVVMV-UHFFFAOYSA-L |
| IUPAC Name | disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate |
| SMILES | [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18602415
|
N/A | 5 g | Amber Glass Bottle |
Each for $261.29
|
|
||||
|
18602416
|
N/A | 25 g | Amber Glass Bottle |
Each for $640.04
|
|
||||
|
18602418
|
N/A | 100 g | Amber Glass Bottle |
Each for $1,156.79
|
|
||||
|
18602417
|
N/A | 1 kg | Amber Glass Bottle |
Each for $2,062.25
|
|
||||
Description
Spectrum™ Chemical Eosin Y, Reagent, ACS , a form of eosin, is used as an acidic red stain for indicating cytoplasm material. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Chemical Identifiers
| 17372-87-1 | |
| 691.86 | |
| disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate |
| C20H6Br4Na2O5 | |
| SEACYXSIPDVVMV-UHFFFAOYSA-L | |
| [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12 |
Specifications
| Crystalline Powder | |
| 100% | |
| C20H6Br4Na2O5 | |
| [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12 | |
| 691.86 | |
| Eosin Y |
| 17372-87-1 | |
| Red to Brown | |
| SEACYXSIPDVVMV-UHFFFAOYSA-L | |
| disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate | |
| ACS |