missing translation for 'onlineSavingsMsg'
Learn More

Eosin Y, Reagent, ACS, Spectrum™ Chemical

Catalog No. 18602415
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602415 5 g Amber Glass Bottle
18602416 25 g Amber Glass Bottle
18602418 100 g Amber Glass Bottle
18602417 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602415 Supplier Spectrum Chemical Mfg Cor Supplier No. E12955GM
Get Up to $500 in Chemicals with Purchase
View all promotions

E1295, 17372-87-1, C20H6Br4Na2O5

Spectrum™ Chemical Eosin Y, Reagent, ACS , a form of eosin, is used as an acidic red stain for indicating cytoplasm material. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 17372-87-1
Molecular Formula C20H6Br4Na2O5
Molecular Weight (g/mol) 691.86
InChI Key SEACYXSIPDVVMV-UHFFFAOYSA-L
IUPAC Name disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12

Specifications

Physical Form Crystalline Powder
CAS Max % 100%
Packaging Amber Glass Bottle
Color Red to Brown
Quantity 5 g
Grade ACS
Chemical Name or Material Eosin Y