missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enrofloxacin 98.0+%, TCI America™
$138.61 - $415.80
Chemical Identifiers
| CAS | 93106-60-6 |
|---|---|
| Molecular Formula | C19H22FN3O3 |
| Molecular Weight (g/mol) | 359.40 |
| MDL Number | MFCD00792463 |
| InChI Key | SPFYMRJSYKOXGV-UHFFFAOYSA-N |
| Synonym | 1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid |
| PubChem CID | 71188 |
| ChEBI | CHEBI:35720 |
| IUPAC Name | 1-cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid |
| SMILES | CCN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1)C(O)=O |
Chemical Identifiers
| 93106-60-6 | |
| 359.40 | |
| SPFYMRJSYKOXGV-UHFFFAOYSA-N | |
| 71188 | |
| 1-cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid |
| C19H22FN3O3 | |
| MFCD00792463 | |
| 1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid | |
| CHEBI:35720 | |
| CCN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1)C(O)=O |
Specifications
| Enrofloxacin | |
| 93106-60-6 | |
| Yellow | |
| MFCD00792463 | |
| SPFYMRJSYKOXGV-UHFFFAOYSA-N | |
| 1-cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid | |
| 71188 | |
| 359.40 | |
| Enrofloxacin |
| Crystalline Powder | |
| 225°C | |
| C19H22FN3O3 | |
| 1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid | |
| CCN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1)C(O)=O | |
| 359.40 | |
| CHEBI:35720 | |
| ≥98.0% (HPLC,T) |
Safety and Handling
RTECSNumber : VB1993650
TSCA : No