missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enrofloxacin 98.0+%, TCI America™
Supplier: TCI America E078625G
| Quantity | 25 g |
|---|
Chemical Identifiers
| 93106-60-6 | |
| 359.40 | |
| SPFYMRJSYKOXGV-UHFFFAOYSA-N | |
| 71188 | |
| 1-cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid |
| C19H22FN3O3 | |
| MFCD00792463 | |
| 1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid | |
| CHEBI:35720 | |
| CCN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1)C(O)=O |
Specifications
| Enrofloxacin | |
| Crystalline Powder | |
| 225°C | |
| 25 g | |
| MFCD00792463 | |
| SPFYMRJSYKOXGV-UHFFFAOYSA-N | |
| 1-cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid | |
| 71188 | |
| 359.40 |
| Enrofloxacin | |
| 93106-60-6 | |
| Yellow | |
| C19H22FN3O3 | |
| 1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid | |
| CCN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1)C(O)=O | |
| 359.40 | |
| CHEBI:35720 | |
| ≥98.0% (HPLC,T) |
Safety and Handling
RTECSNumber : VB1993650
TSCA : No