missing translation for 'onlineSavingsMsg'
Learn More

Enrofloxacin 98.0+%, TCI America™

Catalog No. E07865G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07865G 5 g
E078625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07865G Supplier TCI America Supplier No. E07865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 93106-60-6
Molecular Formula C19H22FN3O3
Molecular Weight (g/mol) 359.40
MDL Number MFCD00792463
InChI Key SPFYMRJSYKOXGV-UHFFFAOYSA-N
Synonym 1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid
PubChem CID 71188
ChEBI CHEBI:35720
IUPAC Name 1-cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
SMILES CCN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1)C(O)=O

Specifications

Type Enrofloxacin
Physical Form Crystalline Powder
Melting Point 225°C
Color Yellow
Quantity 5 g
Formula Weight 359.40
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Enrofloxacin
RTECSNumber VB1993650
TSCA No