missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENPP5 Recombinant Rabbit Monoclonal Antibody (23GB5700)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553101
Description
ENPP5 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
This gene encodes a type-I transmembrane glycoprotein. Studies in rat suggest the encoded protein may play a role in neuronal cell communications. Alternatively spliced transcript variants have been described.Chemical Identifiers
| ENPP5 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| ENPP5 | |
| AW061004; D17Abb1e; EC 3.1; ectonucleotide pyrophosphatase/phosphodiesterase 5; ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative); ectonucleotide pyrophosphatase/phosphodiesterase family member 5; E-NPP 5; ENPP5; E-NPP5; KIAA0879; mKIAA4152; Npp5; NPP-5; UNQ550/PRO1107 | |
| ENPP5 | |
| Primary | |
| Affinity chromatography |
| 23GB5700 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P84039, Q9EQG7, Q9UJA9 | |
| 316249, 59084, 83965 | |
| A synthesized peptide derived from human ENPP5 (400-477AA). | |
| Antibody |
Specifications
| ENPP5 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ENPP5 | |
| AW061004; D17Abb1e; EC 3.1; ectonucleotide pyrophosphatase/phosphodiesterase 5; ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative); ectonucleotide pyrophosphatase/phosphodiesterase family member 5; E-NPP 5; ENPP5; E-NPP5; KIAA0879; mKIAA4152; Npp5; NPP-5; UNQ550/PRO1107 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 316249, 59084, 83965 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5700 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P84039, Q9EQG7, Q9UJA9 | |
| ENPP5 | |
| A synthesized peptide derived from human ENPP5 (400-477AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |