missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ENPP5 Recombinant Rabbit Monoclonal Antibody (23GB5700)
GREENER_CHOICE

Catalog No. p-200055960
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553100 100 μL
MA553101 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553100 Supplier Invitrogen™ Supplier No. MA553100
Only null left

Rabbit Recombinant Monoclonal Antibody

ENPP5 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes a type-I transmembrane glycoprotein. Studies in rat suggest the encoded protein may play a role in neuronal cell communications. Alternatively spliced transcript variants have been described.

Specifications

Antigen ENPP5
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5700
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ENPP5
Gene Accession No. P84039, Q9EQG7, Q9UJA9
Gene Alias AW061004; D17Abb1e; EC 3.1; ectonucleotide pyrophosphatase/phosphodiesterase 5; ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative); ectonucleotide pyrophosphatase/phosphodiesterase family member 5; E-NPP 5; ENPP5; E-NPP5; KIAA0879; mKIAA4152; Npp5; NPP-5; UNQ550/PRO1107
Gene Symbols ENPP5
Host Species Rabbit
Immunogen A synthesized peptide derived from human ENPP5 (400-477AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 316249, 59084, 83965
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less