missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Endoproteinase Glu-C, Excision Grade, Staphylococcus aureus

Catalog No. 32471350UG
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32471350UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32471350UG Supplier MilliporeSigma™ Supplier No. 32471350UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Serine protease that specifically hydrolyzes peptide bonds at carboxylic side of glutamic acid and aspartic acid residues

  • Native endoproteinase Glu-C from Staphylococcus aureus
  • Inhibited by DFP and α2-macroglobulin
  • Suggested working concentration: 1:20 to 1:100 (protease:protein by weight) for sequence analysis

Specifications

CAS 66676-43-5
Content And Storage 2°C to 8°C
Form Lyophilized
Quantity 50 μg
Synonym V8 Protease
Solubility Water
Product Type Endoproteinase