missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Endoproteinase Asp-N, Excision Grade, Pseudomonas fragi

Catalog No. 3247082UG
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3247082UG 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3247082UG Supplier MilliporeSigma™ Supplier No. 3247082UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for protein sequencing or sequence verification, analysis of protein structural domains, and cleavage of fusion proteins

  • Native endoproteinase Asp-N from Pseudomonas fragi
  • Metalloprotease that specifically hydrolyzes proteins at the N-terminal side of aspartic acid and cysteic acid residues
  • Inhibited by aprotinin, DFP, leupeptin, and TLCK
  • Suggested working concentration: 1:20 to 1:100 (protease:protein by weight) for sequence analysis

Specifications

Formulation Lyophilized from 10mM Tris-HCl, pH 7.5.
Purity ≥90% by SDS-PAGE
CAS 9001-92-7
Content And Storage 2°C to 8°C
Form Lyophilized Solid
Quantity 2 μg
Solubility Water
Product Type Endoproteinase