missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Endopeptidase Lys-C, Achromobacter lyticus

Catalog No. 3247962U
Change view
Click to view available options
Quantity:
2 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3247962U 2 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3247962U Supplier MilliporeSigma™ Supplier No. 3247962U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifically hydrolyzes amide and peptide ester bonds at carboxylic side of lysine and S-aminoethylcysteine residues

  • Native endopeptidase Lys-C from Achromobacter lyticus
  • Lysyl endopeptidase is a serine protease secreted by soil bacteria

Specifications

CAS 78642-25-8
Content And Storage -20°C
Form Lyophilized
Quantity 2 U
Solubility Tris-HCl Buffer, Water
Product Type Endopeptidase