missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Endonuclease IV, E.coli (2 U/μL)

Catalog No. FEREN0591
Change view
Click to view available options
Quantity:
100 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREN0591 100 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEREN0591 Supplier Thermo Scientific™ Supplier No. EN0591

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cut oxidative damaged DNA at AP sites with this enzyme, which recognizes apurinic/apyrimidinic sites and cleaves the phosphodiester bond 5' to the lesion.

Endonuclease IV (Endo IV) recognizes apurinic/apyrimidinic (AP) sites of dsDNA and cleaves the phosphodiester bond 5' to the lesion generating a hydroxyl group at the 3'-terminus. The enzyme can also act as a 3'-diesterase that is able to release 3'-phosphoglycolate or 3'-phosphate from the damaged ends of dsDNA . Endo IV possesses also a 3'≥5' exonuclease activity. Its progression on substrates is sensitive to ionic strength, metal ions, EDTA, and reducing conditions. Substrates with 3'-recessed ends are preferred substrates for the 3'≥5' exonuclease activity .

The enzyme has no requirement for Mg2+ and is fully active in the presence of EDTA in moderate concentrations.

Applications
  • Studies of DNA damage and repair
  • Single cell electrophoresis (comet assay)
  • Antitumor drug research
  • DNA structure research
  • SNP analysis

Specifications

Concentration 2 U/μL
Enzyme Endonuclease
Quantity 100 units
Product Type Endonuclease IV

For Research Use Only. Not for use in diagnostic procedures.