missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Empty Glass GC Column, for Agilent™ 5880, 5890, 6890, 7820, and 7890 (Configuration A)

Catalog No. 111016215
Change view
Click to view available options
Length (Metric):
1 m
1.8 m
2 m
Diameter (Metric) Inner:
2 mm
4 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Inner
111016215 1 m 2 mm
111004386 1.8 m 2 mm
111004356 2 m 2 mm
111020823 1.8 m 4 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 111016215 Supplier MilliporeSigma™ Supelco™ Supplier No. 21203U

Empty column may be used for packing adsorbents for temperature-programmed desorption (TPD) instrumentation.

Silane treated empty glass column. Fittings are not included. This column fits Agilent 5880, 5890, 6890, 7820, and 7890 (configuration A, on-column injection, all detectors except TCD).

Specifications

For Use With (Application) Gas Chromatography (GC)
Length (Metric) 1 m
Material Glass Column
Packing Material None
Type Empty GC
Diameter (Metric) Inner 2 mm