missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Upstate Bovine Pituitary Extract
Designed for use as a cell culture media supplement
$543.00 - $786.40
Specifications
| Content And Storage | Stable for 2 years at 4°C from date of shipment. Rehydrated: Stable for 3 months at -20°C |
|---|---|
| For Use With (Application) | Cell Culture |
Specifications
| Stable for 2 years at 4°C from date of shipment. Rehydrated: Stable for 3 months at -20°C | |
| Cell Culture |