missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Ultrafiltration Membranes - YM

Catalog No. 14442AM
Change view
Click to view available options
Diameter (Metric):
2.5 cm
4.45 cm
6.35 cm
7.6 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
14442AM 7.6 cm
14422AM 4.45 cm
14432 6.35 cm
14412 2.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14442AM Supplier MilliporeSigma™ Supplier No. 14442AM

High-recovery membranes for stirred cells

Specifications

Filter Type Adsorptive
Material (Membrane) Regenerated Cellulose
MWCO 100 kDa
Filter Code YM-100
Filtration Area 41.8 cm2
For Use With (Application) Protein purification, protein concentration, protein desalting
Quantity 10 Pack
Type Ultrafiltration Disc
Diameter (English) 2.99 in.
Diameter (Metric) 7.6 cm