missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Ultrafiltration Discs, 30 kDa NMW, Ultracel™ Regenerated Cellulose

Catalog No. pltk04310
Change view
Click to view available options
Diameter (Metric) Filter:
2.5 cm
4.45 cm
4.7 cm
6.35 cm
7.6 cm
9 cm
15 cm
Filtration Area:
13.4 cm2
17.3 cm2
177 cm2
28.7 cm2
4.1 cm2
41.8 cm2
63 cm2
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Filter Filtration Area
PLTK04310 4.45 cm 13.4 cm2
PLTK07610 7.6 cm 41.8 cm2
PLTK09005 9 cm 63 cm2
PLTK02510 2.5 cm 4.1 cm2
PLTK04710 4.7 cm 17.3 cm2
PLTK06210 6.35 cm 28.7 cm2
PLTK15005 15 cm 177 cm2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. PLTK04310 Supplier MilliporeSigma™ Supplier No. PLTK04310
Only null left

30 kDa NMWL, Ultracel™ regenerated cellulose

Assemble your Amicon Stirred Cell with the membrane of your choice. Ultrafiltration cut disc membranes are available in Ultracel™ regenerated cellulose and Biomax™ polyethersulvone (PES). The membranes are available in a range of different pore sizes & diameters.Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic, tight microstructure of Ultracel membranes assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.Microfiltration Durapore™ PVDF cut discs are available 0.1 μm, 0.22 μm, and 0.45 μm pore sizes in both 63.5 mm and 75 mm diameters.Features & Benefits:•Disc diameters include: 25, 44.5, 47, 63.5, 76, 90 and 150 mm •Both Ultracel™, Durapore™, and Biomax™ membranes are available in a wide range of pore sizes to meet your separation requirements •Compatible with Amicon™ Stirred Cells Applications:Concentration, Buffer Exchange, Desalting

Specifications

Filter Type Ultrafiltration
Material (Membrane) Regenerated Cellulose
MWCO 30 kDa
Filter Code PLTK
Filtration Area 13.4 cm2
For Use With (Application) Protein purification; protein concentration; protein desalting
Housing Material Regenerated Cellulose (RC)
Quantity 10Pack
Type Ultrafiltration Disc
Diameter (Metric) Filter 4.45 cm