missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ SynergyPak™ 3 Cartridge for Synergy™ Ultrapure Water Systems

Catalog No. SYPK0SIX3
Change view
Click to view available options
Purification pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
SYPK0SIX3 Purification pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYPK0SIX3 Supplier MilliporeSigma™ Supplier No. SYPK0SIX3

Produces ultrapure water from Deionized (DI) water

  • Baed on “Ion-exchange”, “Activated carbon”, “Depth filtration” technology
  • Integrate all main purification technologies
  • “Plug-and-use” design for easy and rapid replacement

Specifications

Filter Type Ion exchange, Activated carbon, depth filtration technology
For Use With (Application) Produces ultrapure water from DI water