missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ SNAP i.d. 2.0™ Protein Detection Systems for Western Blotting

Catalog No. SNAP2MB2
Change view
Click to view available options
Type:
Midi and MultiBlot
Mini and MultiBlot
Dimensions (L x W x H):
7.5 x 8.4 cm and 4.5 x 8.4 cm
8.5 x 13.5 cm and 4.5 x 8.4 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Dimensions (L x W x H)
SNAP2MB2 Midi and MultiBlot 8.5 x 13.5 cm and 4.5 x 8.4 cm
SNAP2MB1 Mini and MultiBlot 7.5 x 8.4 cm and 4.5 x 8.4 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SNAP2MB2 Supplier MilliporeSigma™ Supplier No. SNAP2MB2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

SNAP i.d. 2.0™ base, holding Frames, two antibody collection trays, vacuum tubing, rolling pad and blot roller, two wetting trays and quick start guide

Detection system with unique vacuum-driven technology and built-in flow distributor that actively drives reagents through membrane

  • System produces blots of high quality
  • For use on immunoblots
  • Detection Method: Chemiluminescence Chromogenic Fluorescence
  • Mini and MultiBlot: 7.5 x 8.4cm and 4.5 x 8.4cm
  • Midi and MultiBlot: 8.5 x 13.5cm and 4.5 x 8.4cm

Western Blotting

Specifications

For Use With (Application) Western Blot
Type Midi and MultiBlot
Includes SNAP i.d. 2.0™ base, holding Frames, two antibody collection trays, vacuum tubing, rolling pad and blot roller, two wetting trays and quick start guide
Quantity 1/Pk.
Dimensions (L x W x H) 8.5 x 13.5 cm and 4.5 x 8.4 cm