Learn More
MilliporeSigma™ SeQuant™ ZIC™-pHILIC Polymeric Analytical PEEK Guard Columns
PEEK coated column
Supplier: MilliporeSigma™ 1504370001
Description
The polymeric SeQuant™ ZIC™-pHILIC columns is designed for particularly difficult separations of polar and hydrophilic compounds. This guard column protects your analytical separation column from contamination.Specifications
| 2.1 mm | |
| 5 μm | |
| 200 AÌ | |
| Guard Column |
| 20 mm | |
| 200 Å | |
| 1/Pk. |