Learn More
MilliporeSigma™ SeQuant™ ZIC-HILIC HPLC Metal-free PEEK Guard Columns and Kit
PEEK coated HPLC column
Supplier: MilliporeSigma™ 1504360001
Description
SeQuant™ ZIC™-HILIC is the ideal choice for separation of polar and hydrophilic compounds. This guard column protects your analytical separation column from contamination.Specifications
| 2.1 mm | |
| 5 μm | |
| 200 AÌ | |
| HPLC Guard Column w/Coupler |
| 20 mm | |
| 200 Å | |
| 3/Pk. |