Learn More
MilliporeSigma™ SeQuant™ ZIC™-cHILIC Metal-free HPLC PEEK Columns
PEEK coated HPLC column
Supplier:  MilliporeSigma™ 1506580001
 View more versions of this product
Catalog No. M1506580001
Only null left
Explore available promotions
Description
Description
SeQuant™ ZIC™-cHILIC is designed for improved separation of polar hydrophilic compounds that have proved difficult to separate by reversed-phase HPLC or previous types of HILIC columns. The orientation of its zwitterionic functional group provides more accessible positive charges, which results in a complementary selectivity with more retention particularly for organic acids and other negatively charged compounds. Especially suitable for LC-MS thanks to an extraordinary high column stability and a minimized column bleed.
Specifications
Specifications
2.1 mm
150 mm
1/Pk.
Column
Product Suggestions
Product Suggestions