Learn More
MilliporeSigma™ Samplicity™ Filtration System Vial Trays
Vacuum filtration system for sample preparation for HPLC
$137.00 - $186.00
Specifications
| For Use With (Equipment) | Samplicity G2 Filtration System |
|---|---|
| Sterility | Non-sterile |
| Catalog Number | Mfr. No. | Type | Material | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Material | Price | Quantity & Availability | ||||
|
09719400
|
MilliporeSigma™
SAMVIALTR |
Vial Trays | Polypropylene |
Pack of 2 Each for $186.00
|
|
||||
|
09719401
|
MilliporeSigma™
SAMWASTTR |
Waste Trays | Recycled Polyester |
Pack of 2 Each for $137.00
|
|
||||
Description
Overview of Samplicity™ G2 System- Samplicity™ G2 Filtration System enables vacuum driven filtration of up to 8 samples at once, directly into HPLC vials.
- Filtration for HPLC sample prep without syringes. Sample is added to an adapter funnel on a standard Millex™ 33 mm filter (Millex™ Filters for the Samplicity™ G2 System) or an integrated funnel and filter (Millex Samplicity™ filter)
- Samplicity™ G2 System has lower ergonomic impact than manual syringe filtration
Applications: HPLC; Filtration of viscous or particulate laden samples for HPLC
Specifications
| Samplicity G2 Filtration System | |
| Non-sterile |