Learn More
MilliporeSigma™ Samplicity™ Filtration System Vial Trays
Catalog No. 09719400
Change view
Click to view available options
Type:
Vial Trays
Waste Trays
Material:
Polypropylene
Recycled Polyester
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Material
09719400 Vial Trays Polypropylene
09719401 Waste Trays Recycled Polyester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09719400 Supplier MilliporeSigma™ Supplier No. SAMVIALTR
Vacuum filtration system for sample preparation for HPLC
Overview of Samplicity™ G2 System
  • Samplicity™ G2 Filtration System enables vacuum driven filtration of up to 8 samples at once, directly into HPLC vials.
  • Filtration for HPLC sample prep without syringes. Sample is added to an adapter funnel on a standard Millex™ 33 mm filter (Millex™ Filters for the Samplicity™ G2 System) or an integrated funnel and filter (Millex Samplicity™ filter)
  • Samplicity™ G2 System has lower ergonomic impact than manual syringe filtration
Applications: HPLC; Filtration of viscous or particulate laden samples for HPLC
Specifications
For Use With (Equipment) Samplicity G2 Filtration System
Material Polypropylene
Sterility Non-sterile
For Use With (Application) Analytical sample prep, High Performance Liquid Chromatography (HPLC)
No. per Pack 2 Trays
Type Vial Trays