missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ RNase A

Catalog No. LSKPMRN30
Change view
Click to view available options
Quantity:
0.9 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LSKPMRN30 0.9 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LSKPMRN30 Supplier MilliporeSigma™ Supplier No. LSKPMRN30

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

As supplied with Montage™ Plasmid MiniprepHTS 96 Kit

Specifications

For Use With (Application) Used as a reagent with montage plasmid miniprepHTS 96 kit for high-purity plasmid or Bacterial Artificial Chromosome minipreps
Content And Storage 15°C to 30°C
Quantity 0.9 mL
Product Type Ribonuclease A