missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ PureProteome™ Magnetic Beads

Catalog No. MILSKMAGN01
Change view
Click to view available options
Quantity:
1 x 0.5 mL
4 x 0.5 mL
Product Type:
NHS flexiBind
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
MILSKMAGN01 1 x 0.5 mL NHS flexiBind
MILSKMAGN04 4 x 0.5 mL NHS flexiBind
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MILSKMAGN01 Supplier MilliporeSigma™ Supplier No. LSKMAGN01

For rapid and reproducible isolation of proteins

Custom affinity bead generation for binding to target ligand

  • Advanced chemistry combined with high surface area provides more binding sites for proteins
  • Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
  • Beads are immobilized in seconds
  • Increased kinetics of bead-protein binding enables shorter incubations
  • Significantly more affordable then competitive magnetic beads

Specifications

Product Type NHS flexiBind
Quantity 1 x 0.5 mL
Particle Size 10 μm
Content And Storage 2° to 8°C
For Use With (Application) Immunoprecipitations, purifying Nucleic Acids, isolating cells/organelles, Protein-protein interactions
Matrix Magnetic Beads