missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ PureProteome™ Lambda Ig Binder Magnetic Beads

Catalog No. LSKMAGLM02
Change view
Click to view available options
Quantity:
2 x 1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LSKMAGLM02 2 x 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LSKMAGLM02 Supplier MilliporeSigma™ Supplier No. LSKMAGLM02

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Requires

PureProteome™ Magnetic Stand (8 well) (Cat # LSKMAGS08); Not delivered.

PureProteome™ magnetic beads are a powerful system to isolate antibodies faster, easier and with better reproducibly then ever before.

The PureProteome™ Lambda Ig Binder Magnetic Bead System is a powerful system that helps researchers purify all Immunoglobulins (i.e IgG, IgA, IgM, IgE and IgD) with high specificity. Researchers can also purify/seperate the Fab portion from the Fc portion of Immunoglobulins.

  • High capacity: Advanced chemistry combined with high surface area provides more binding sites for proteins
  • Consistent results with no sample loss: Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
  • Fast processing time: Beads are immobilized in seconds. Increased kinetics of bead-protein binding enables shorter incubations
  • Economical: Significantly more affordable then competitive magnetic beads

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Applications:

Antibody Purification

Specifications

Product Type Lambda Ig binder Magnetic bead
Quantity 2 x 1mL
Particle Size 10μm
Content And Storage 2° to 8°C
For Use With (Application) Purifying all types of Igs Antibodies
Matrix Magnetic Beads