Learn More
MilliporeSigma™ PureProteome™ Kappa Ig Binder Magnetic Beads
PureProteome magnetic beads are a powerful system to isolate antibodies faster, easier and with better reproducibly then ever before.
Supplier: MilliporeSigma™ LSKMAGKP02
Requires : PureProteome Magnetic Stand (8 well) (Cat # LSKMAGS08)
Description
The PureProteome Kappa Ig Binder Magnetic Bead System is a powerful system that helps researchers purify all Immunoglobulins (i.e IgG, IgA, IgM, IgE and IgD) with high specificity. Researchers can also purify/seperate the Fab portion from the Fc portion of Immunoglobulins.
- High capacity: Advanced chemistry combined with high surface area provides more binding sites for proteins
- Consistent results with no sample loss: Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
- Fast processing time: Beads are immobilized in seconds. Increased kinetics of bead-protein binding enables shorter incubations
- Economical: Significantly more affordable then competitive magnetic beads
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Applications:
Antibody Purification
Specifications
| kappa Ig binder Magnetic bead | |
| 10μm | |
| Antibody purification |
| 2 x 1mL | |
| 2° to 8°C | |
| Magnetic Beads |