missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ PureProteome™ Carboxy FlexiBind Magnetic Bead System
For rapid and reproducible isolation of proteins
$1,750.00 - $1,750.00
Specifications
| Product Type | Carboxy flexiBind Magnetic bead system |
|---|---|
| Content And Storage | 2° to 8°C |
| For Use With (Application) | Immunoprecipitations, purifying Nucleic Acids, isolating cells/organelles, Protein-protein interactions |
| dimMatrix | Magnetic Beads |
| Catalog Number | Mfr. No. | Quantity | Particle Size | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Particle Size | Price | Quantity | ||||
|
LSKMAG03C10
|
MilliporeSigma™
LSKMAG03CBX10 |
10 mL | 0.3μm |
N/A
|
|
||||
Description
Custom affinity bead generation for binding to target ligand.
- Advanced chemistry combined with high surface area provides more binding sites for proteins
- Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
- Beads are immobilized in seconds
- Increased kinetics of bead-protein binding enables shorter incubations
- Significantly more affordable then competitive magnetic beads
Research Use Only
Specifications
| Carboxy flexiBind Magnetic bead system | |
| 2° to 8°C | |
| Immunoprecipitations, purifying Nucleic Acids, isolating cells/organelles, Protein-protein interactions | |
| Magnetic Beads |