missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ PureProteome™ Carboxy FlexiBind Magnetic Bead System

Catalog No. LSKMAG03C10
Change view
Click to view available options
Quantity:
10 mL
Particle Size:
0.3μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Particle Size
LSKMAG03C10 10 mL 0.3μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LSKMAG03C10 Supplier MilliporeSigma™ Supplier No. LSKMAG03CBX10

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For rapid and reproducible isolation of proteins

Custom affinity bead generation for binding to target ligand.

  • Advanced chemistry combined with high surface area provides more binding sites for proteins
  • Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
  • Beads are immobilized in seconds
  • Increased kinetics of bead-protein binding enables shorter incubations
  • Significantly more affordable then competitive magnetic beads

Research Use Only

Specifications

Product Type Carboxy flexiBind Magnetic bead system
Quantity 10 mL
Particle Size 0.3μm
Content And Storage 2° to 8°C
For Use With (Application) Immunoprecipitations, purifying Nucleic Acids, isolating cells/organelles, Protein-protein interactions
Matrix Magnetic Beads