missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ PureProteome™ Albumin Magnetic Beads

Catalog No. LSKMAGL10
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LSKMAGL10 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LSKMAGL10 Supplier MilliporeSigma™ Supplier No. LSKMAGL10

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Faster, easier serum depletion with greater insight into your sample

  • Beads are conjugated to an antibody specific for human serum albumin
  • Provides rapid, scalable, and reproducible means to deplete >98% of albumin from serum and plasma samples, facilitating the detection and analysis of proteins of interest
  • Adjust bead volume based on sample volume and total protein concentration
  • No manipulation of tubes or multipart assemblies required
  • No need for centrifugation; magnetic field immobilizes beads in seconds
  • Particle Size: 10μm

Specifications

Product Type Albumin Magnetic bead
Quantity 10 mL
Particle Size 10μm
Content And Storage 2° to 8°C
For Use With (Application) Serum depletion
Matrix Magnetic Beads