Learn More
MilliporeSigma™ pT7Blue Blunt Vector (linearized)
$388.00 - $592.00
Specifications
Promoter T7
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
701743
View Documents Promotion Details
MilliporeSigma™
701743
20 Reactions
Each for $388.00
Only null left
701744
View Documents Promotion Details
MilliporeSigma™
701744
40 Reactions
Each for $592.00
Only null left
Description
Description
pT7Blue contains the pUC19 backbone (including high-copy number origin of replication and lac sequences), a T7 promoter, f1 origin of replication, and modified multiple cloning region. The multiple cloning region contains an EcoR V site used for T-cloning flanked by an Nde I site, which allows inserts to be conveniently subcloned into the Nde I site of many pET vectors.
Specifications
Specifications
T7
Store at -20°C. Avoid freeze/thaw cycles.