Learn More
MilliporeSigma™ pT7Blue Blunt Vector (linearized)
Catalog No. 701743
Change view
Click to view available options
Quantity:
20 Reactions
40 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
701743 20 Reactions
701744 40 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 701743 Supplier MilliporeSigma™ Supplier No. 701743
pT7Blue contains the pUC19 backbone (including high-copy number origin of replication and lac sequences), a T7 promoter, f1 origin of replication, and modified multiple cloning region. The multiple cloning region contains an EcoR V site used for T-cloning flanked by an Nde I site, which allows inserts to be conveniently subcloned into the Nde I site of many pET vectors.
Specifications
Promoter T7
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Quantity 20 Reactions
For Research Use Only