Learn More
MilliporeSigma™ pSTBlue-1 Blunt Vector (linearized)
$388.00 - $592.00
Specifications
| Promoter | Dual Opposed T7/SP6 |
|---|---|
| Content And Storage | Store at -70°C. Avoid freeze/thaw cycles. |
Description
The pSTBlue-1 vector is a multi-purpose cloning vector featuring a versatile multiple cloning region, blue/white screening, dual opposed T7/SP6 promoters and dual kanamycin/ampicillin resistance. Restriction sites producing 4-base 3' overhangs are conveniently positioned near each end of the polylinker to facilitate the generation of unidirectional deletions using exonuclease III.Specifications
| Dual Opposed T7/SP6 | |
| Store at -70°C. Avoid freeze/thaw cycles. |
For Research Use Only