Learn More
MilliporeSigma™ pSTBlue-1 Blunt Vector (linearized)
Supplier: MilliporeSigma™ 701883
Description
The pSTBlue-1 vector is a multi-purpose cloning vector featuring a versatile multiple cloning region, blue/white screening, dual opposed T7/SP6 promoters and dual kanamycin/ampicillin resistance. Restriction sites producing 4-base 3' overhangs are conveniently positioned near each end of the polylinker to facilitate the generation of unidirectional deletions using exonuclease IIIChemical Identifiers
| Dual Opposed T7/SP6 | |
| Store at -70°C. Avoid freeze/thaw cycles. |
Specifications
| Dual Opposed T7/SP6 | |
| Store at -70°C. Avoid freeze/thaw cycles. | |
| 20 Reactions |
For Research Use Only