missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ PetriSlide™ Dishes Preloaded with Absorbent Pads

Catalog No. PDMA04700
Change view
Click to view available options
Diameter (Metric):
47 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
PDMA04700 47 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PDMA04700 Supplier MilliporeSigma™ Supplier No. PDMA04700
Edge

Substitute for Petri Dishes. PetriSlide™ Dish Preloaded with Absorbent Pads is designed to hold filter securely in place. Transparent cover allows microscopic examination without removal. Rectangular base has rounded corners for mounting on microscope stage.

Key Applications:
  • Clinical Lab
  • Food and Beverage
  • Industrial QC
  • Environmental Analysis

Specifications

Color Clear
Material Polystyrene
Shape Round
Sterility Sterile
Type Petri Dish
For Use With (Application) For microbiological analysis
Diameter (Metric) 47 mm