missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ PetriSlide™ for Contamination Analysis

Catalog No. PD1504700
Change view
Click to view available options
Diameter (Metric):
47 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
PD1504700 47 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PD1504700 Supplier MilliporeSigma™ Supplier No. PD1504700
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Substitute for Petri Dishes. PetriSlide™ for Contamination Analysis includes a transparent cover allowing microscopic examination without removal. Rectangular base has rounded corners for mounting on microscope stage.

Substitute for Petri Dishes. Merck Millipore™ PetriSlide™ for Contamination Analysis includes a transparent cover allowing microscopic examination without removal. Rectangular base has rounded corners for mounting on microscope stage.

Polystyrene storage and examination containers for culturing microorganisms.

  • Hold 47mm filter securely in place
  • Transparent cover allows microscopic examination without removal
  • Rectangular base has rounded corners for mounting on microscopic stage
  • Stored in cartons or standard slide cases
  • Use pure cellulose pads for microbiological analysis
  • Use without pads for contamination analysis

 

Specifications

Closure Color Transparent
Color Clear
Material Polystyrene
Shape Round
Sterility Non-sterile
Type PetriSlide Container
For Use With (Application) For use without pads for contamination analysis
Diameter (Metric) 47 mm