missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Petri Dishes and Petri-Pad™ Dishes

Available with or without absorbent pads

Supplier:  MilliporeSigma™ PD2004700

 View more versions of this product

Catalog No. PD2004700


Only null left

Requires : When using with liquid medium, add sterile absorbent pads (order Cat. No. AP10 045 S0, 47mm Sterile Pads; or AP10 045 S1, Millipore Pad Dispenser with Sterile Pads)

Description

Description

Available with or without absorbent pads

Petri Dishes

  • 47mm sterilized Petri dishes
  • Use with solid agar medium
  • Dishes are made of molded polystyrene
Petri-Pad Dishes
  • Presterilized disposable 47mm dishes preloaded with sterilized absorbent pads
  • Eliminate labor and cost of hand-loading pads and minimize contamination
  • Pads are pure cellulose

Specifications

Chemical Identifiers

Single-use
Sterile
47 mm
Polystyrene
Sleeve

Specifications

Single-use
Sterile
For solid agar medium or liquid medium with sterile absorbent pads
100 Pk. (5 Sleeves)
Polystyrene
Dish without Pad
Sleeve
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions