missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Pellicon™ XL Devices

Catalog No. PXB100C50
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
1000 kDa
30 kDa
300 kDa
5 kDa
50 kDa
500 kDa
8 kDa
pH Range:
1 to 14
2 to 21
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO pH Range
PXB100C50 100 kDa 1 to 14
PXB005A50 5 kDa 1 to 14
PXB01MC50 1000 kDa 1 to 14
PXB008A50 8 kDa 1 to 14
PXB050A50 50 kDa 1 to 14
PXB300C50 300 kDa 1 to 14
PXB500C50 500 kDa 1 to 14
PXC01MC50 1000 kDa 2 to 21
PXC005C50 5 kDa 2 to 21
PXC030C50 30 kDa 2 to 21
PXC300C50 300 kDa 2 to 21
PXC010C50 10 kDa 2 to 21
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. PXB100C50 Supplier MilliporeSigma™ Supplier No. PXB100C50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Operating instructions and Certificate of Quality

Small-volume tangential flow filter device

Designed for process development and sample preparation, with an optimal processing volume range of 50 to 1000mL.

  • True linear scalability with same flow path, channel length, and channel height as other established Pellicon products
  • Can operate with peristaltic pump or Labscale™ TFF system Robust thermoplastic (polypropylene) construction
  • Controlled channel height
  • Choice of Biomax™ PES (polyethersulfone) or Ultracel™ PLC (composite regenerated cellulose) membranes

Concentration and desalting of proteins, buffer exchange, depyrogenation, clarification, and preparation of material for clinical trials.

Order Info

Packaged one per box.

Specifications

Filter Type Ultrafiltration
Format Cassette
Material Polypropylene
MWCO 100 kDa
Pore Size 100 μm
Filtration Area 50 cm2
For Use With (Application) Protein purification, analytical sample preparation
Includes Operating instructions and certificate of quality
Max. Inlet Pressure 80 psi at 50°C
pH Range 1 to 14
Quantity 1 Pack
Recirculation Rate 0.03 to 0.05L/min.
Type Pellicon XL Ultrafiltration Device
Volume (English) Working 0.5 oz.
Volume (Metric) Hold Up 3.2 mL
Volume (Metric) Working 15 mL
Dimensions (L x W) 7.4 x 1.2 in. (18.8 x 3cm)
Length (English) 7.4 in.
Length (Metric) 18.8 cm
Width (English) 1.2 in.
Width (Metric) 3 cm
Show More Show Less