Learn More
MilliporeSigma™ Novagen™ BL21(DE3) Singles™ Competent Cells
BL21(DE3) is a chemically competent E. coli cell suitable for transformation and high level protein expression using a T7 RNA polymerase-IPTG induction system.
$281.02 - $484.60
Specifications
Efficiency Guaranteed efficiency: >2 x 107 cfu/μg
Content And Storage ≤-70°C
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
702354
View Documents
MilliporeSigma™
702354
22 Reactions
Each for $484.60
Only null left
702353
View Documents
MilliporeSigma™
702353
11 Reactions
Each for $281.02
Only null left
Description
Description
BL21 strains are the most widely used hosts for protein expression from pET recombinants and have the advantage of being deficient in both lon and ompT proteases.
Specifications
Specifications
Guaranteed efficiency: >2 x 107 cfu/μg
≤-70°C
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'