Learn More
MilliporeSigma™ Novagen™ BL21(DE3) Singles™ Competent Cells
Catalog No. 702354
Change view
Click to view available options
Quantity:
11 Reactions
22 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
702354 22 Reactions
702353 11 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 702354 Supplier MilliporeSigma™ Supplier No. 702354
BL21(DE3) is a chemically competent E. coli cell suitable for transformation and high level protein expression using a T7 RNA polymerase-IPTG induction system.
BL21 strains are the most widely used hosts for protein expression from pET recombinants and have the advantage of being deficient in both lon and ompT proteases.
Specifications
Efficiency Guaranteed efficiency: >2 x 107 cfu/μg
Content And Storage ≤-70°C
Quantity 22 Reactions