missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Immobilon™-PSQ PVDF Membranes

For Western blots and protein sequencing

Supplier:  MilliporeSigma™ ISEQ08130

 View more versions of this product

Catalog No. ISEQ08130


Only null left

Description

Description

Immobilon-PSQ Membrane:
  • 0.2 μm pore size and a large internal structure
  • Higher protein adsorption and sequencing yields than other membranes
  • Recommended for blotting proteins less than 20 kDa
  • Prevents blow-through of low molecular weight proteins
  • Specifications

    Chemical Identifiers

    0.2 μm

    Specifications

    8.5 cm
    For sequencing low-abundance or low-molecular weight (<20,000 Daltons) proteins
    Immobilon-PSQ
    8.5 x 13.5 cm
    13.5 cm
    Sheet
    0.2 μm
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Promotions

    Promotions