missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ HumanKine™ Human Thermostable bFGF Recombinant Protein, Xeno-Free

$229.00 - $864.00

Specifications

Name Human Thermostable bFGF Recombinant Protein, Xeno-Free
Source Human 293 protein expression system
Recombinant Recombinant
Purity or Quality Grade Endotoxin free and greater than 95% pure by SDS-PAGE
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
GF44650UG
View Documents
MilliporeSigma™
GF44650UG
50 μg
Each for $864.00
Only null left
 
GF44610UG
View Documents
MilliporeSigma™
GF44610UG
10 μg
Each for $229.00
Only null left
 
Description

Description

Specifications

Specifications

Human Thermostable bFGF Recombinant Protein, Xeno-Free
Human 293 protein expression system
Recombinant
Endotoxin free and greater than 95% pure by SDS-PAGE
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'